Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plxdc1
Entrez Gene ID
72324
Gene Name
plexin domain containing 1
Gene Aliases
2410003I07Rik, AI848450, Tem7
Chromosome Location
Chr.11: 97923237 - 97986667 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163608.1 NP_001157080.1 3 - 4 553 72
NM_028199.3 NP_082475.3 3 - 4 532 72
GenBank mRNA AF378760.1 - 3 - 4 466 72
AK010361.1 - 3 - 4 532 72
AK036144.1 - 3 - 4 507 72
AK153684.1 - 3 - 4 519 72
BC114352.2 - 3 - 4 452 72

View Full Product Details