The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029624.4 | NP_083900.3 | 8 - 9 | 1258 | 75 |
NR_045520.1 | 6 - 7 | 937 | 75 | ||
GenBank mRNA | AK010312.1 | - | 8 - 9 | 1229 | 75 |
AK041659.1 | - | 6 - 7 | 925 | 75 | |
AK154622.1 | - | 8 - 9 | 1255 | 75 | |
AK169983.1 | - | 8 - 9 | 1256 | 75 | |
BC020104.1 | - | 8 - 9 | 1265 | 75 |