The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029624.4 | NP_083900.3 | 10 - 11 | 1549 | 79 |
NR_045520.1 | 8 - 9 | 1228 | 79 | ||
GenBank mRNA | AK010312.1 | - | 10 - 11 | 1520 | 79 |
AK041659.1 | - | 8 - 9 | 1216 | 79 | |
AK154622.1 | - | 10 - 11 | 1546 | 79 | |
AK169983.1 | - | 10 - 11 | 1547 | 79 | |
BC020104.1 | - | 10 - 11 | 1556 | 79 |