The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109684.1 | NP_001103154.1 | 1 - 2 | 590 | 90 |
NM_001109685.1 | NP_001103155.1 | 2 - 3 | 608 | 90 | |
NM_144531.3 | NP_653114.3 | 1 - 2 | 590 | 90 | |
GenBank mRNA | AK052809.1 | - | 2 - 3 | 609 | 90 |
AK173090.1 | - | 1 - 2 | 590 | 90 | |
BC022941.1 | - | 1 - 2 | 594 | 90 | |
CD349067.1 | - | 1 - 2 | 165 | 90 |