The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007964.2 | NP_031990.2 | 1 - 2 | 122 | 68 |
GenBank mRNA | BI737814.1 | - | 1 - 2 | 46 | 68 |
U53586.1 | - | 1 - 2 | 122 | 68 |