The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008391.4 | NP_032417.3 | 7 - 8 | 895 | 82 |
GenBank mRNA | AK087926.1 | - | 6 - 7 | 763 | 82 |
AK132099.1 | - | 7 - 8 | 908 | 82 | |
AK155025.1 | - | 7 - 8 | 880 | 82 | |
BC006577.1 | - | 7 - 8 | 888 | 82 | |
BC110666.1 | - | 7 - 8 | 867 | 82 | |
J03168.1 | - | 7 - 8 | 850 | 82 |