Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Baz1b
Entrez Gene ID
22385
Gene Name
bromodomain adjacent to zinc finger domain, 1B
Gene Aliases
C87820, WSTF, Wbscr9
Chromosome Location
Chr.5: 135187323 - 135246129 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011714.2 NP_035844.2 13 - 14 3762 94
GenBank mRNA AF084480.1 - 13 - 14 3721 94
AK037737.1 - 7 - 8 970 94
BC141399.1 - 13 - 14 3665 94
BC171975.1 - 13 - 14 3664 94

View Full Product Details