The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018869.3 | NP_061357.3 | 14 - 15 | 2367 | 113 |
GenBank mRNA | AF040746.1 | - | 14 - 15 | 1777 | 113 |
AK135076.1 | - | 14 - 15 | 2369 | 113 | |
AK156465.1 | - | 13 - 14 | 2743 | 113 | |
AK161821.1 | - | 14 - 15 | 1847 | 113 | |
BC019379.1 | - | 14 - 15 | 1640 | 113 |