Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fktn
Entrez Gene ID
246179
Gene Name
fukutin
Gene Aliases
D830030O17Rik, Fcmd
Chromosome Location
Chr.4: 53714107 - 53765785 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139309.4 NP_647470.1 7 - 8 1159 120
GenBank mRNA AB077383.1 - 8 - 9 1343 120
AJ511807.1 - 8 - 9 1179 120
AK052357.1 - 10 - 11 1588 120
AK085931.1 - 7 - 8 1160 120
AK142744.1 - - 1319 120
AK144743.1 - 7 - 8 960 120
BC017538.1 - 8 - 9 1175 120

View Full Product Details