Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Yipf3
Entrez Gene ID
28064
Gene Name
Yip1 domain family, member 3
Gene Aliases
C6orf109, D17Wsu94e, KLIP1
Chromosome Location
Chr.17: 46248070 - 46252548 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145353.2 NP_663328.2 2 - 3 450 62
GenBank mRNA AF197931.1 - 1 - 2 210 62
AK042644.1 - 1 - 2 122 62
AK149957.1 - 2 - 3 283 62
BC019384.1 - 2 - 3 431 62
BC100575.1 - 2 - 3 382 62

View Full Product Details