The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021516.4 | NP_067491.2 | 12 - 13 | 1803 | 66 |
NM_022801.4 | NP_073712.2 | 12 - 13 | 1803 | 66 | |
GenBank mRNA | AF240782.1 | - | 12 - 13 | 1348 | 66 |
AF240783.1 | - | 12 - 13 | 1348 | 66 | |
AK002654.1 | - | 3 - 4 | 365 | 66 | |
AK075742.1 | - | 3 - 4 | 365 | 66 | |
AK132247.1 | - | 12 - 13 | 1787 | 66 | |
AK166157.1 | - | 12 - 13 | 1792 | 66 | |
BC026445.1 | - | 5 - 6 | 708 | 66 | |
BC066071.1 | - | 11 - 12 | 2438 | 66 | |
CA575239.1 | - | 2 - 3 | 194 | 66 |