Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tnrc6a
Entrez Gene ID
233833
Gene Name
trinucleotide repeat containing 6a
Gene Aliases
2010321I05Rik, 3110054G10Rik, AW557223, CAGH26, D130023A07Rik, GW182, Tnrc6
Chromosome Location
Chr.7: 123031101 - 123195296 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144925.3 NP_659174.3 17 - 18 4923 69
GenBank mRNA AK044679.1 - 5 - 6 1307 69
AK138426.1 - 7 - 8 973 69
AK142808.1 - 2 - 3 292 69
AK142809.1 - 2 - 3 292 69
AK147327.1 - 17 - 18 4628 69
BC005741.1 - 6 - 7 758 69

View Full Product Details