The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285997.1 | NP_001272926.1 | 1 - 2 | 141 | 76 |
NM_001285998.1 | NP_001272927.1 | 1 - 2 | 141 | 76 | |
NM_145150.3 | NP_660132.2 | 1 - 2 | 141 | 76 | |
GenBank mRNA | AK029140.1 | - | 1 - 2 | 128 | 76 |
AK081506.1 | - | 1 - 2 | 114 | 76 | |
BC005475.1 | - | 1 - 2 | 75 | 76 | |
BC059808.1 | - | 1 - 2 | 86 | 76 | |
CF736005.1 | - | 1 - 2 | 141 | 76 |