Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tm4sf4
Entrez Gene ID
229302
Gene Name
transmembrane 4 superfamily member 4
Gene Aliases
Iltmp
Chromosome Location
Chr.3: 57425410 - 57441677 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145539.2 NP_663514.2 1 - 2 381 73
GenBank mRNA AK148484.1 - 1 - 2 349 73
AK165731.1 - 1 - 2 349 73
BC010814.1 - 1 - 2 332 73

View Full Product Details