The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146097.3 | NP_666209.1 | 4 - 5 | 512 | 100 |
NR_033744.1 | 4 - 5 | 579 | 100 | ||
GenBank mRNA | AK076027.1 | - | 4 - 5 | 467 | 100 |
AK132291.1 | - | 4 - 5 | 498 | 100 | |
BC018472.1 | - | 4 - 5 | 454 | 100 | |
BC053745.1 | - | 4 - 5 | 547 | 100 |