Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hoxb2
Entrez Gene ID
103889
Gene Name
homeobox B2
Gene Aliases
AI894218, Hox-2.8, Hoxbes2
Chromosome Location
Chr.11: 96351632 - 96354020 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134032.2 NP_598793.2 2 - 2 969 101
GenBank mRNA AB221540.1 - 1 - 1 215 101
BC019414.1 - 1 - 1 554 101
BC138494.1 - 2 - 2 888 101
BC138495.1 - 2 - 2 888 101

View Full Product Details