The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172124.2 | NP_742122.2 | 1 - 2 | 1304 | 65 |
GenBank mRNA | AB055902.1 | - | 1 - 2 | 1250 | 65 |
AK048146.1 | - | 1 - 2 | 1220 | 65 | |
AK052640.1 | - | 1 - 2 | 1304 | 65 | |
AK136293.1 | - | 1 - 2 | 225 | 65 | |
AK220433.1 | - | - | 1212 | 65 | |
BC056368.1 | - | 1 - 2 | 1196 | 65 | |
BC058082.1 | - | 1 - 2 | 1196 | 65 | |
BC092280.1 | - | 2 - 3 | 969 | 65 |