Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chdh
Entrez Gene ID
218865
Gene Name
choline dehydrogenase
Gene Aliases
AU041472, D630034H06Rik
Chromosome Location
Chr.14: 30009000 - 30040466 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136240.1 NP_001129712.1 5 - 6 1411 86
NM_172264.2 NP_758468.2 5 - 6 1209 86
NM_175343.5 NP_780552.1 5 - 6 1384 86
GenBank mRNA AK030900.1 - 5 - 6 1385 86
AK087570.1 - 5 - 6 1320 86
AK164042.1 - 5 - 6 1247 86
BC039186.1 - 5 - 6 1209 86

View Full Product Details