Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prrc1
Entrez Gene ID
73137
Gene Name
proline-rich coiled-coil 1
Gene Aliases
1190002C06Rik, 2310058D16Rik, 3110038B19Rik, 9430085A19Rik
Chromosome Location
Chr.18: 57354733 - 57392956 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028447.3 NP_082723.2 6 - 7 1098 64
GenBank mRNA AK004443.1 - 6 - 7 1098 64
AK143537.1 - 6 - 7 1090 64
BC014817.1 - 6 - 7 1014 64

View Full Product Details