Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Unk
Entrez Gene ID
217331
Gene Name
unkempt homolog (Drosophila)
Gene Aliases
B230379M23Rik, Zc3h5, Zc3hdc5, mKIAA1753
Chromosome Location
Chr.11: 116030295 - 116061214 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286006.1 NP_001272935.1 6 - 7 949 59
NM_172569.4 NP_766157.1 6 - 7 949 59
GenBank mRNA AK046395.1 - 6 - 7 944 59
AK078095.1 - 6 - 7 940 59
AK129438.1 - 6 - 7 927 59
AK134361.1 - 6 - 7 1069 59
BC003195.1 - 3 - 4 395 59
BC039964.1 - 6 - 7 911 59
BC054452.1 - 6 - 7 886 59

View Full Product Details