The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172739.4 | NP_766327.3 | 7 - 7 | 8195 | 134 |
GenBank mRNA | AK032559.1 | - | 1 - 1 | 2948 | 134 |
AK136766.1 | - | 6 - 6 | 4871 | 134 | |
AK137250.1 | - | - | 243 | 134 |