The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172879.3 | NP_766467.2 | 3 - 4 | 2431 | 85 |
GenBank mRNA | AK029981.1 | - | 3 - 4 | 2432 | 85 |
BC141026.1 | - | 3 - 4 | 2258 | 85 | |
BC145170.1 | - | 3 - 4 | 2258 | 85 |