Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd3e
Entrez Gene ID
12501
Gene Name
CD3 antigen, epsilon polypeptide
Gene Aliases
AI504783, CD3, CD3epsilon, T3e
Chromosome Location
Chr.9: 44998735 - 45009662 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007648.4 NP_031674.1 2 - 3 128 110
GenBank mRNA BC145926.1 - 2 - 3 128 110
BC145954.1 - 2 - 3 128 110
J02990.1 - 2 - 3 129 110

View Full Product Details