Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nfkbiz
Entrez Gene ID
80859
Gene Name
nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, zeta
Gene Aliases
AA408868, INAP, Mail
Chromosome Location
Chr.16: 55811377 - 55838641 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159394.1 NP_001152866.1 8 - 9 1767 70
NM_001159395.1 NP_001152867.1 9 - 10 1649 70
NM_030612.3 NP_085115.1 10 - 11 1997 70
GenBank mRNA AB020974.1 - 10 - 11 2209 70
AB026551.2 - 8 - 9 1767 70
AB047549.2 - 9 - 10 1662 70
AK082908.1 - 8 - 9 1766 70
AK136509.1 - 5 - 6 1236 70
AK141318.1 - 8 - 9 1768 70
AK149946.1 - 9 - 10 1645 70
AK150519.1 - 8 - 9 1766 70
AK152016.1 - 8 - 9 1768 70
AK156826.1 - 8 - 9 1767 70
AK159093.1 - 8 - 9 1768 70
AK159459.1 - 8 - 9 1768 70
AK159645.1 - 8 - 9 1768 70
AK160065.1 - 8 - 9 1766 70
BC058188.1 - 8 - 9 1752 70

View Full Product Details