Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnaic1
Entrez Gene ID
68922
Gene Name
dynein, axonemal, intermediate chain 1
Gene Aliases
1110066F04Rik, BB124644, Dnai1, b2b1526Clo
Chromosome Location
Chr.4: 41569663 - 41638158 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175138.4 NP_780347.2 8 - 9 882 71
GenBank mRNA AK030213.1 - 8 - 9 816 71
AK047808.1 - 8 - 9 883 71
BC138785.1 - 8 - 9 827 71
BC138787.1 - 8 - 9 827 71

View Full Product Details