Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Actbl2
Entrez Gene ID
238880
Gene Name
actin, beta-like 2
Gene Aliases
4732495G21Rik
Chromosome Location
Chr.13: 111255013 - 111257749 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175497.3 NP_780706.1 1 - 1 2559 144
GenBank mRNA AK029110.1 - 1 - 1 2560 144

View Full Product Details