The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177333.3 | NP_796307.2 | 12 - 13 | 2160 | 85 |
GenBank mRNA | AK088333.1 | - | 12 - 13 | 2154 | 85 |
AK089979.1 | - | 12 - 13 | 2170 | 85 | |
AK131141.1 | - | 12 - 13 | 2144 | 85 | |
AK141537.1 | - | 12 - 13 | 2160 | 85 | |
BC031743.1 | - | 12 - 13 | 2148 | 85 |