Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dpy19l3
Entrez Gene ID
233115
Gene Name
dpy-19-like 3 (C. elegans)
Gene Aliases
6030410G08, 9330164H19Rik
Chromosome Location
Chr.7: 35685500 - 35754454 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178704.3 NP_848819.2 18 - 19 2225 69
GenBank mRNA AF538322.1 - 18 - 19 2209 69
AK031345.1 - 18 - 19 2217 69
AK033395.1 - 18 - 19 2182 69
AK034217.1 - 18 - 19 2226 69
AK045758.1 - 18 - 19 2126 69
BC050065.1 - 18 - 19 2169 69
BC059897.1 - 18 - 19 2121 69

View Full Product Details