Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Triml1
Entrez Gene ID
244448
Gene Name
tripartite motif family-like 1
Gene Aliases
4933403D14, BC050188
Chromosome Location
Chr.8: 43129806 - 43180391 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177742.4 NP_808410.1 2 - 3 613 70
GenBank mRNA AK077092.1 - 2 - 3 615 70
BC141202.1 - 2 - 3 592 70

View Full Product Details