The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301173.1 | NP_001288102.1 | 2 - 3 | 345 | 60 |
NM_177791.3 | NP_808459.1 | 2 - 3 | 345 | 60 | |
GenBank mRNA | AK076644.1 | - | 2 - 3 | 337 | 60 |
BC092530.1 | - | 2 - 3 | 345 | 60 | |
BC125431.1 | - | 2 - 3 | 337 | 60 | |
BC125433.1 | - | 2 - 3 | 337 | 60 |