The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010658.3 | NP_034788.1 | 1 - 1 | 2301 | 122 |
GenBank mRNA | AK154830.1 | - | 1 - 1 | 1663 | 122 |
BC038256.1 | - | 1 - 1 | 2249 | 122 |