Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mtus1
Entrez Gene ID
102103
Gene Name
mitochondrial tumor suppressor 1
Gene Aliases
AI481402, ATBP135, Atip1, B430010I23Rik, B430305I03Rik, C85752, Cctsg1-440, MD44, MTSG1, mKIAA1288
Chromosome Location
Chr.8: 40990912 - 41133983 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF173380.1 - 2 - 3 516 70
AF493235.1 - 2 - 3 581 70
AK129324.1 - 2 - 3 663 70
AY246699.1 - 2 - 3 339 70
AY626781.1 - 2 - 3 674 70
BC041777.1 - 5 - 6 1723 70
BC042206.1 - 5 - 6 1723 70

View Full Product Details