The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026675.2 | NP_080951.1 | 5 - 6 | 817 | 65 |
GenBank mRNA | AK002252.1 | - | 5 - 6 | 818 | 65 |
BC019768.1 | - | 4 - 5 | 794 | 65 |