The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009336.2 | NP_033362.2 | 3 - 4 | 442 | 92 |
GenBank mRNA | AK140284.1 | - | 3 - 4 | 440 | 92 |
AK155191.1 | - | 3 - 4 | 440 | 92 | |
BC004834.1 | - | 3 - 4 | 420 | 92 | |
BC043029.1 | - | 3 - 4 | 443 | 92 | |
D43643.1 | - | 3 - 4 | 413 | 92 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AK137145.1 | - | Chr. - - [-] on Build | 92 |