The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013491.2 | NP_038519.1 | 20 - 21 | 2509 | 106 |
GenBank mRNA | AK036735.1 | - | 20 - 21 | 2511 | 106 |
AY046403.1 | - | 20 - 21 | 2334 | 106 | |
AY046404.1 | - | 21 - 22 | 2413 | 106 | |
BC114336.1 | - | 20 - 21 | 2474 | 106 |