Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cryz
Entrez Gene ID
12972
Gene Name
crystallin, zeta
Gene Aliases
Sez9
Chromosome Location
Chr.3: 154596711 - 154623182 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009968.3 NP_034098.1 3 - 4 855 101
GenBank mRNA AK098082.1 - 3 - 4 339 101
AK133820.1 - 3 - 4 856 101
AK134281.1 - 3 - 4 340 101
BC043076.1 - 3 - 4 316 101
S70056.1 - 3 - 4 367 101

View Full Product Details