Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gdi1
Entrez Gene ID
14567
Gene Name
guanosine diphosphate (GDP) dissociation inhibitor 1
Gene Aliases
GDIA, GDIalpha
Chromosome Location
Chr.X: 74305012 - 74311867 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010273.4 NP_034403.1 5 - 6 967 56
GenBank mRNA AK028880.1 - 5 - 6 724 56
AK156224.1 - 5 - 6 735 56
AK171216.1 - 5 - 6 724 56
BC013758.1 - 5 - 6 648 56
BC025606.1 - 5 - 6 734 56
BC037598.1 - 5 - 6 701 56

View Full Product Details