Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gdi1
Entrez Gene ID
14567
Gene Name
guanosine diphosphate (GDP) dissociation inhibitor 1
Gene Aliases
GDIA, GDIalpha
Chromosome Location
Chr.X: 74305012 - 74311867 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010273.4 NP_034403.1 8 - 9 1372 77
GenBank mRNA AF251042.1 - 8 - 9 1041 77
AK028880.1 - 8 - 9 1129 77
AK032869.1 - 7 - 8 1378 77
AK156224.1 - 8 - 9 1140 77
AK171216.1 - 8 - 9 1129 77
BC013758.1 - 8 - 9 1053 77
BC025606.1 - 8 - 9 1139 77
BC037598.1 - 8 - 9 1106 77
U07950.1 - 4 - 5 615 77

View Full Product Details