Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lck
Entrez Gene ID
16818
Gene Name
lymphocyte protein tyrosine kinase
Gene Aliases
Hck-3, Lsk, Lskt, p56<lck>, p56Lck
Chromosome Location
Chr.4: 129548344 - 129573641 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162432.1 NP_001155904.1 4 - 5 370 100
NM_001162433.1 NP_001155905.1 4 - 5 412 100
NM_010693.3 NP_034823.1 3 - 4 467 100
GenBank mRNA AK088001.1 - 3 - 4 468 100
AK134013.1 - 3 - 4 457 100
BC011474.1 - 4 - 5 377 100
BQ563042.1 - 4 - 5 361 100
CA577097.1 - 1 - 2 103 100
M12056.1 - 3 - 4 443 100
X03533.1 - 3 - 4 473 100

View Full Product Details