Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fyb
Entrez Gene ID
23880
Gene Name
FYN binding protein
Gene Aliases
ADAP, B630013F22Rik, FYB-120/130, Slap130, p120/130, p120/p130
Chromosome Location
Chr.15: 6522797 - 6665608 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278269.1 NP_001265198.1 16 - 17 2404 90
NM_011815.5 NP_035945.1 17 - 18 2542 90
GenBank mRNA AF001863.1 - 16 - 17 2380 90
AF061744.1 - 17 - 18 2518 90
AK137907.1 - 16 - 17 2360 90
AK161411.1 - 3 - 4 410 90
AK172164.1 - 10 - 11 774 90
BC141352.1 - 17 - 18 2483 90

View Full Product Details