Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C8b
Entrez Gene ID
110382
Gene Name
complement component 8, beta polypeptide
Gene Aliases
4930439B20Rik, AI595927
Chromosome Location
Chr.4: 104764576 - 104804549 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133882.2 NP_598643.1 2 - 3 322 66
GenBank mRNA AB077304.1 - 2 - 3 244 66
AB077305.1 - 2 - 3 244 66
AB077306.1 - 2 - 3 244 66
AK050313.1 - 2 - 3 323 66
BC096382.1 - 2 - 3 296 66

View Full Product Details