Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc41
Entrez Gene ID
230654
Gene Name
leucine rich repeat containing 41
Gene Aliases
AA409966, AW555107, D630045E04Rik, D730026A16Rik, MUF1
Chromosome Location
Chr.4: 116075465 - 116097043 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK028000.1 - 2 - 3 303 104
AK052762.1 - 5 - 6 1606 104
AK154613.1 - 5 - 6 1752 104
AK184800.1 - 1 - 2 105 104
AK185861.1 - 1 - 2 104
AK210637.1 - 1 - 2 105 104
BC005765.1 - 4 - 5 1473 104
BC007169.1 - 2 - 3 631 104
BC022976.1 - 5 - 6 1684 104
BC024940.1 - 5 - 6 1681 104
BC071258.1 - 5 - 6 1617 104

View Full Product Details