Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc17a8
Entrez Gene ID
216227
Gene Name
solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 8
Gene Aliases
BC042593, Vglut3
Chromosome Location
Chr.10: 89574020 - 89621249 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182959.3 NP_892004.1 11 - 12 1820 72
GenBank mRNA AF510321.1 - 11 - 12 1491 72
AK082743.1 - 11 - 12 1569 72
AK149687.1 - 9 - 10 1223 72
BC042593.1 - 11 - 12 1835 72

View Full Product Details