The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172099.1 | NP_001165570.1 | 2 - 3 | 738 | 87 |
NM_198013.3 | NP_932130.2 | 3 - 4 | 936 | 87 | |
GenBank mRNA | AK049250.1 | - | 3 - 4 | 900 | 87 |
AK162604.1 | - | 2 - 3 | 318 | 87 | |
BC024476.1 | - | 3 - 4 | 785 | 87 | |
BC025434.1 | - | 3 - 4 | 790 | 87 | |
BC029708.1 | - | 3 - 4 | 869 | 87 | |
BC131989.1 | - | 2 - 3 | 709 | 87 | |
BC131991.1 | - | 2 - 3 | 709 | 87 | |
BC145206.1 | - | 2 - 3 | 709 | 87 | |
CK332869.1 | - | 2 - 3 | 182 | 87 |