The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146370.1 | NP_666482.1 | 1 - 1 | 918 | 85 |
GenBank mRNA | BC120658.1 | - | 1 - 1 | 950 | 85 |
BC120660.1 | - | 1 - 1 | 950 | 85 |