The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102474.1 | NP_001095944.1 | 8 - 9 | 1217 | 102 |
NM_177578.4 | NP_808246.1 | 6 - 7 | 1177 | 102 | |
GenBank mRNA | AK040383.1 | - | 6 - 7 | 1178 | 102 |
EF494894.1 | - | 8 - 9 | 1217 | 102 | |
EF494895.1 | - | 6 - 7 | 1001 | 102 | |
EF494896.1 | - | 7 - 8 | 869 | 102 |