Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chic2
Entrez Gene ID
74277
Gene Name
cysteine-rich hydrophobic domain 2
Gene Aliases
1700081B18Rik, 4930502K01Rik, BTL
Chromosome Location
Chr.5: 75006424 - 75044626 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK006962.1 - 1 - 2 445 145
AK172289.1 - 1 - 2 438 145
BC048435.1 - 2 - 3 198 145
BC116971.1 - 1 - 2 255 145
BC116973.1 - 1 - 2 255 145

View Full Product Details