The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011188.3 | NP_035318.1 | 12 - 13 | 1540 | 61 |
GenBank mRNA | AK005083.1 | - | 11 - 12 | 1163 | 61 |
AK076874.1 | - | 12 - 13 | 1542 | 61 | |
AK146919.1 | - | 11 - 12 | 1181 | 61 | |
AK153413.1 | - | 11 - 12 | 1184 | 61 | |
AK168548.1 | - | 11 - 12 | 1181 | 61 | |
BC005462.1 | - | 11 - 12 | 1190 | 61 |