Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ebf3
Entrez Gene ID
13593
Gene Name
early B cell factor 3
Gene Aliases
3110018A08Rik, O/E-2, mKIAA4201
Chromosome Location
Chr.7: 137193671 - 137322810 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113414.1 NP_001106885.1 3 - 4 889 64
NM_001113415.1 NP_001106886.1 3 - 4 889 64
NM_010096.3 NP_034226.1 3 - 4 889 64
GenBank mRNA AK220542.1 - 3 - 4 889 64
BC067018.1 - 4 - 5 644 64
U82442.1 - 2 - 3 186 64
U92702.1 - 3 - 4 430 64
U92704.1 - 3 - 4 430 64

View Full Product Details