Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ebf3
Entrez Gene ID
13593
Gene Name
early B cell factor 3
Gene Aliases
3110018A08Rik, O/E-2, mKIAA4201
Chromosome Location
Chr.7: 137193671 - 137322810 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113414.1 NP_001106885.1 9 - 10 1411 113
NM_001113415.1 NP_001106886.1 9 - 10 1438 113
NM_010096.3 NP_034226.1 9 - 10 1411 113
GenBank mRNA AK138602.1 - 4 - 5 600 113
AK220542.1 - 9 - 10 1411 113
BC067018.1 - 10 - 11 1166 113
CF727275.1 - 3 - 4 303 113
U92702.1 - 9 - 10 979 113
U92704.1 - 9 - 10 952 113

View Full Product Details